Please login to your online account to display your discounted pricing

Rabbit anti-Melan-A/MART-1, Clone: A19-P, Novus Biologicals™

$478.29 - $478.29

Specifications

Antigen Melan-A/MART-1
Clone A19-P
Immunogen Peptide derived from C-terminal sequence of human Melan A.
Gene Accession No. Q16655
Gene ID (Entrez) 2315
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP13015105

 
novus biologicals canada
NBP13015105ML
0.5mL Each for $478.29
Description & Specifications

Specifications

Antigen Melan-A/MART-1
Clone A19-P
Immunogen Peptide derived from C-terminal sequence of human Melan A.
Gene Accession No. Q16655
Gene ID (Entrez) 2315
Formulation 20mM Tris-HCl, pH 8.0 with 20mg/mL BSA.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rabbit
Isotype IgG
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)