Please login to your online account to display your discounted pricing

Rabbit anti-Melan-A/MART-1, Clone: A19-P, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP13015105ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP13015105

  • / Each
 

Description & Specifications

Specifications

Host Species Rabbit
Immunogen Peptide derived from C-terminal sequence of human Melan A.
Format Affinity Purified
Gene Accession No. Q16655
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Quantity 0.5mL
Storage Requirements Store at 4°C. Do not freeze.
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 2315
Conjugate Unlabeled
Formulation 20mM Tris-HCl, pH 8.0 with 20mg/mL BSA.
Isotype IgG
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody is specific for the C terminal portions of Melan-A
Clone A19-P
Cross Reactivity Human
Cross Reactivity Mouse
Antigen Melan-A/MART-1
Purification Method Affinity Purified