Please login to your online account to display your discounted pricing

Rabbit anti-MCM2, Clone: SP85, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP21249005ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP21249005

  • / Each
 

Description & Specifications

Specifications

Antigen MCM2
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Clone SP85
Conjugate Unlabeled
Format Purified
Formulation PBS/1% BSA buffer pH 7.6
Host Species Rabbit
Immunogen Synthetic peptide derived from internal region of human MCM2 protein.
Isotype IgG
Purification Method Purified
Quantity 0.5mL
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 4171
Test Specificity Predicted to cross react with Bovine, Canine, Frog, Mouse, Rat.
Reconstitution Protein A purified
Target Species Human