Please login to your online account to display your discounted pricing

Rabbit anti-MCM2, Clone: SP85, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP21249001ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP21249001

  • / Each
 

Description & Specifications

Specifications

Antigen MCM2
Format Purified
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Immunogen Synthetic peptide derived from internal region of human MCM2 protein.
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Host Species Rabbit
Purification Method Purified
Quantity 0.1mL
Storage Requirements Store at 4°C. Do not freeze.
Gene ID (Entrez) 4171
Reconstitution Protein A purified
Formulation PBS/1% BSA buffer pH 7.6
Isotype IgG
Test Specificity Predicted to cross react with Bovine, Canine, Frog, Mouse, Rat.
Clone SP85