Please login to your online account to display your discounted pricing

Rabbit anti-Luteinizing Hormone, Clone: SP132, Novus Biologicals™

$309.05 - $551.93

Specifications

Antigen Luteinizing Hormone
Clone SP132
Immunogen Recombinant human LH protein
Formulation PBS/1% BSA buffer pH 7.6
Monoclonal or Polyclonal Monoclonal
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP21251005

 
novus biologicals canada
NBP21251005ML
0.5mL Each for $551.93

NBP21251001

 
novus biologicals canada
NBP21251001ML
0.1mL Each for $309.05
Description & Specifications

Specifications

Antigen Luteinizing Hormone
Clone SP132
Immunogen Recombinant human LH protein
Formulation PBS/1% BSA buffer pH 7.6
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rabbit
Cross Reactivity Human
Isotype IgG
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)