Please login to your online account to display your discounted pricing

Rabbit anti-Luteinizing Hormone, Clone: SP132, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP21251005ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP21251005

  • / Each
 

Description & Specifications

Specifications

Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Formulation PBS/1% BSA buffer pH 7.6
Host Species Rabbit
Isotype IgG
Quantity 0.5mL
Primary or Secondary Primary
Immunogen Recombinant human LH protein
Antigen Luteinizing Hormone
Monoclonal or Polyclonal Monoclonal
Regulatory Status RUO
Conjugate Unlabeled
Storage Requirements Store at 4°C. Do not freeze.
Purification Method Purified
Clone SP132
Test Specificity Predicted to cross react with Mk.
Reconstitution Protein A purified
Format Purified
Cross Reactivity Human