Please login to your online account to display your discounted pricing

Rabbit anti-Luteinizing Hormone, Clone: SP132, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP21251001ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP21251001

  • / Each
 

Description & Specifications

Specifications

Conjugate Unlabeled
Test Specificity Predicted to cross react with Mk.
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Cross Reactivity Human
Format Purified
Immunogen Recombinant human LH protein
Isotype IgG
Formulation PBS/1% BSA buffer pH 7.6
Antigen Luteinizing Hormone
Clone SP132
Purification Method Purified
Reconstitution Protein A purified
Regulatory Status RUO
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Host Species Rabbit
Storage Requirements Store at 4°C. Do not freeze.
Quantity 0.1mL