Please login to your online account to display your discounted pricing

Rabbit anti-Lambda light chain, Clone: K22-Y, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP221837200UL

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP22183720

  • / Each
 

Description & Specifications

Specifications

Format Affinity Purified
Formulation 20mM Tris-HCI, pH 8.0 20mg/mL BSA.
Gene Accession No. P01842
Host Species Rabbit
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Purification Method Affinity Purified
Quantity 200μL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Clone K22-Y
Cross Reactivity Human
Antigen Lambda light chain
Immunogen Peptide derived from N-terminal sequence of human lambda light chain IgG.
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Conjugate Unlabeled