Please login to your online account to display your discounted pricing

Rabbit anti-Lambda light chain, Clone: K22-Y, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP13014205ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP13014205

  • / Each
 

Description & Specifications

Specifications

Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Immunogen Peptide derived from N-terminal sequence of human lambda light chain.
Format Affinity Purified
Formulation 20mM Tris-HCl, pH 8.0 with 20mg/mL BSA
Gene Accession No. P01842
Host Species Rabbit
Isotype IgG
Quantity 0.5mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Clone K22-Y
Antigen Lambda light chain
Purification Method Affinity Purified
Test Specificity This antibody is specific for the N terminus of the lambda light chain.
Conjugate Unlabeled
Cross Reactivity Human