Please login to your online account to display your discounted pricing

Rabbit anti-Ku80/XRCC5, Clone: SP130, Novus Biologicals™

$309.05 - $552.33

Specifications

Antigen Ku80/XRCC5
Clone SP130
Immunogen Synthetic peptide corresponding to an internal region of human Ku80 protein.
Gene ID (Entrez) 7520
Formulation PBS/1% BSA buffer pH 7.6
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP21250901

 
novus biologicals canada
NBP21250901ML
0.1mL Each for $309.05

NBP21250905

 
novus biologicals canada
NBP21250905ML
0.5mL Each for $552.33
Description & Specifications

Specifications

Antigen Ku80/XRCC5
Clone SP130
Immunogen Synthetic peptide corresponding to an internal region of human Ku80 protein.
Gene ID (Entrez) 7520
Formulation PBS/1% BSA buffer pH 7.6
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rabbit
Cross Reactivity Human
Isotype IgG
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting