Please login to your online account to display your discounted pricing

Rabbit anti-Ku80/XRCC5, Clone: SP130, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP21250905ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP21250905

  • / Each
 

Description & Specifications

Specifications

Antigen Ku80/XRCC5
Format Purified
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Immunogen Synthetic peptide corresponding to an internal region of human Ku80 protein.
Purification Method Purified
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Formulation PBS/1% BSA buffer pH 7.6
Host Species Rabbit
Isotype IgG
Quantity 0.5mL
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Test Specificity Predicted to cross react with Bovine, Mouse, Rat.
Reconstitution Protein A purified
Clone SP130
Conjugate Unlabeled
Cross Reactivity Human
Gene ID (Entrez) 7520