Please login to your online account to display your discounted pricing

Rabbit anti-Ku80/XRCC5, Clone: SP130, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP21250901ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP21250901

  • / Each
 

Description & Specifications

Specifications

Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Immunogen Synthetic peptide corresponding to an internal region of human Ku80 protein.
Conjugate Unlabeled
Isotype IgG
Cross Reactivity Human
Gene ID (Entrez) 7520
Test Specificity Predicted to cross react with Bovine, Mouse, Rat.
Purification Method Purified
Regulatory Status RUO
Format Purified
Host Species Rabbit
Primary or Secondary Primary
Formulation PBS/1% BSA buffer pH 7.6
Reconstitution Protein A purified
Monoclonal or Polyclonal Monoclonal
Quantity 0.1mL
Storage Requirements Store at 4°C. Do not freeze.
Clone SP130
Antigen Ku80/XRCC5