Please login to your online account to display your discounted pricing

Rabbit anti-Kappa Light Chain, Clone: A21-Y, Novus Biologicals™

$332.66 - $612.00

Specifications

Immunogen Peptide derived from N-terminal sequence of IGKC.
Gene Accession No. P01834
Gene ID (Entrez) 3514
Formulation 20mM Tris-HCl, pH 8.0 with 20mg/mL BSA. Immunogen affinity purified.
Monoclonal or Polyclonal Monoclonal
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP13013901

 
novus biologicals canada
NBP13013901ML
0.1mL Each for $332.66

NBP13013905

 
novus biologicals canada
NBP13013905ML
0.5mL Each for $612.00
Description & Specifications

Specifications

Immunogen Peptide derived from N-terminal sequence of IGKC.
Gene Accession No. P01834
Gene ID (Entrez) 3514
Formulation 20mM Tris-HCl, pH 8.0 with 20mg/mL BSA. Immunogen affinity purified.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Host Species Rabbit
Cross Reactivity Human
Format Affinity Purified
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Clone A21-Y