Please login to your online account to display your discounted pricing

Rabbit anti-Kappa Light Chain, Clone: A21-Y, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP13013905ML

 View more versions of this product

Catalog No. NBP13013905

  • / Each
 

Description & Specifications

Specifications

Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Cross Reactivity Human
Antigen Kappa Light Chain
Format Affinity Purified
Formulation 20mM Tris-HCl, pH 8.0 with 20mg/mL BSA. Immunogen affinity purified.
Immunogen Peptide derived from N-terminal sequence of IGKC.
Isotype IgG
Quantity 0.5mL
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3514
Conjugate Unlabeled
Gene Accession No. P01834
Host Species Rabbit
Regulatory Status RUO
Clone A21-Y