Please login to your online account to display your discounted pricing

Rabbit anti-J Chain, Clone: SP105, Novus Biologicals™

$309.33 - $551.93

Specifications

Antigen J Chain
Clone SP105
Immunogen Synthetic peptide near the internal region of human J-chain.
Formulation PBS/1% BSA buffer pH 7.6
Monoclonal or Polyclonal Monoclonal
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP21250101

 
novus biologicals canada
NBP21250101ML
0.1mL Each for $309.33

NBP21250105

 
novus biologicals canada
NBP21250105ML
0.5mL Each for $551.93
Description & Specifications

Specifications

Antigen J Chain
Clone SP105
Immunogen Synthetic peptide near the internal region of human J-chain.
Formulation PBS/1% BSA buffer pH 7.6
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rabbit
Cross Reactivity Human
Cross Reactivity Mouse
Isotype IgG
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)