Please login to your online account to display your discounted pricing

Rabbit anti-J Chain, Clone: SP105, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP21250105ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP21250105

  • / Each
 

Description & Specifications

Specifications

Conjugate Unlabeled
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Monoclonal or Polyclonal Monoclonal
Host Species Rabbit
Cross Reactivity Human
Cross Reactivity Mouse
Format Purified
Formulation PBS/1% BSA buffer pH 7.6
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Clone SP105
Immunogen Synthetic peptide near the internal region of human J-chain.
Quantity 0.5mL
Regulatory Status RUO
Reconstitution Protein A purified
Isotype IgG
Purification Method Purified
Antigen J Chain