Please login to your online account to display your discounted pricing

Rabbit anti-iNOS, Clone: SP126, Novus Biologicals™

$309.05 - $806.17

Specifications

Antigen iNOS
Clone SP126
Immunogen Synthetic peptide near the C-terminus of mouse iNOS protein.
Gene ID (Entrez) 4843
Formulation PBS/1% BSA buffer pH 7.6 with less than 0.1% sodium azide.
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP17451401

 
novus biologicals canada
NBP17451401ML
0.1mL Each for $309.05

NBP17451405

 
novus biologicals canada
NBP17451405ML
0.5mL Each for $551.93

NBP17451410

 
novus biologicals canada
NBP17451410ML
1mL Each for $806.17
Description & Specifications

Specifications

Antigen iNOS
Clone SP126
Immunogen Synthetic peptide near the C-terminus of mouse iNOS protein.
Gene ID (Entrez) 4843
Formulation PBS/1% BSA buffer pH 7.6 with less than 0.1% sodium azide.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rabbit
Cross Reactivity Human
Isotype IgG
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)