Please login to your online account to display your discounted pricing

Rabbit anti-iNOS, Clone: SP126, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP17451401ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP17451401

  • / Each
 

Description & Specifications

Specifications

Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Format Purified
Immunogen Synthetic peptide near the C-terminus of mouse iNOS protein.
Isotype IgG
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 4843
Conjugate Unlabeled
Antigen iNOS
Formulation PBS/1% BSA buffer pH 7.6 with less than 0.1% sodium azide.
Host Species Rabbit
Purification Method Purified
Quantity 0.1mL
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Reconstitution Protein A purified
Clone SP126
Cross Reactivity Human