Please login to your online account to display your discounted pricing

Rabbit anti-iNOS, Clone: K13-A, Novus Biologicals™

$406.00 - $406.00

Specifications

Antigen iNOS
Clone K13-A
Immunogen Peptide derived from the human iNOS sequence.
Gene Accession No. P35228
Gene ID (Entrez) 4843
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP13378001

 
novus biologicals canada
NBP13378001ML
0.1mL Each for $406.00
Description & Specifications

Specifications

Antigen iNOS
Clone K13-A
Immunogen Peptide derived from the human iNOS sequence.
Gene Accession No. P35228
Gene ID (Entrez) 4843
Formulation 20mM Tris-HCl, pH 8.0, 20mg/mL BSA. Immunogen affinity purified.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Rabbit
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blot