Please login to your online account to display your discounted pricing

Rabbit anti-Ikaros/IKZF1, Clone: SP108, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP21250305ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP21250305

  • / Each
 

Description & Specifications

Specifications

Formulation PBS/1% BSA buffer pH 7.6
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Quantity 0.5mL
Antigen Ikaros/IKZF1
Format Purified
Immunogen Synthetic peptide near the N-terminus of human IKAROS.
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Gene ID (Entrez) 10320
Reconstitution Protein A purified
Conjugate Unlabeled
Cross Reactivity Human
Host Species Rabbit
Isotype IgG
Purification Method Purified
Monoclonal or Polyclonal Monoclonal
Test Specificity Predicted to cross react with Mouse, Rat.
Clone SP108