Please login to your online account to display your discounted pricing

Rabbit anti-Ikaros/IKZF1, Clone: SP108, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP21250301ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP21250301

  • / Each
 

Description & Specifications

Specifications

Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Antigen Ikaros/IKZF1
Formulation PBS/1% BSA buffer pH 7.6
Host Species Rabbit
Immunogen Synthetic peptide near the N-terminus of human IKAROS.
Regulatory Status RUO
Gene ID (Entrez) 10320
Reconstitution Protein A purified
Clone SP108
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Isotype IgG
Purification Method Purified
Quantity 0.1mL
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Test Specificity Predicted to cross react with Mouse, Rat.