Please login to your online account to display your discounted pricing

Rabbit anti-ICOS, Clone: SP98, Novus Biologicals™

$329.00 - $551.93

Specifications

Antigen ICOS
Clone SP98
Immunogen Synthetic peptide corresponding to C-terminus of human CD278.
Gene ID (Entrez) 29851
Formulation PBS/1% BSA buffer pH 7.6
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP21249901

 
novus biologicals canada
NBP21249901ML
0.1mL Each for $329.00

NBP21249905

 
novus biologicals canada
NBP21249905ML
0.5mL Each for $551.93
Description & Specifications

Specifications

Antigen ICOS
Clone SP98
Immunogen Synthetic peptide corresponding to C-terminus of human CD278.
Gene ID (Entrez) 29851
Formulation PBS/1% BSA buffer pH 7.6
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rabbit
Cross Reactivity Human
Isotype IgG
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)