Please login to your online account to display your discounted pricing

Rabbit anti-ICOS, Clone: SP98, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP21249905ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP21249905

  • / Each
 

Description & Specifications

Specifications

Format Purified
Formulation PBS/1% BSA buffer pH 7.6
Cross Reactivity Human
Monoclonal or Polyclonal Monoclonal
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Regulatory Status RUO
Isotype IgG
Quantity 0.5mL
Purification Method Purified
Primary or Secondary Primary
Antigen ICOS
Immunogen Synthetic peptide corresponding to C-terminus of human CD278.
Storage Requirements Store at 4°C. Do not freeze.
Conjugate Unlabeled
Reconstitution Protein A purified
Host Species Rabbit
Clone SP98
Gene ID (Entrez) 29851
Test Specificity Predicted to cross react with Rat.