Please login to your online account to display your discounted pricing

Rabbit anti-ICOS, Clone: SP98, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP21249901ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP21249901

  • / Each
 

Description & Specifications

Specifications

Format Purified
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Cross Reactivity Human
Clone SP98
Isotype IgG
Immunogen Synthetic peptide corresponding to C-terminus of human CD278.
Host Species Rabbit
Antigen ICOS
Reconstitution Protein A purified
Quantity 0.1mL
Gene ID (Entrez) 29851
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Predicted to cross react with Rat.
Formulation PBS/1% BSA buffer pH 7.6
Conjugate Unlabeled
Storage Requirements Store at 4°C. Do not freeze.
Purification Method Purified
Monoclonal or Polyclonal Monoclonal