Please login to your online account to display your discounted pricing

Rabbit anti-HNF-3 alpha/FoxA1, Clone: SP88, Novus Biologicals™

$321.00 - $569.00

Specifications

Antigen HNF-3 alpha/FoxA1
Clone SP88
Immunogen Synthetic peptide derived from near C-terminus of human FOXA1 protein.
Gene ID (Entrez) 3169
Formulation PBS/1% BSA buffer pH 7.6
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP21249301

 
novus biologicals canada
NBP21249301ML
0.1mL Each for $321.00

NBP21249305

 
novus biologicals canada
NBP21249305ML
0.5mL Each for $569.00
Description & Specifications

Specifications

Antigen HNF-3 alpha/FoxA1
Clone SP88
Immunogen Synthetic peptide derived from near C-terminus of human FOXA1 protein.
Gene ID (Entrez) 3169
Formulation PBS/1% BSA buffer pH 7.6
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rabbit
Cross Reactivity Human
Isotype IgG
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)