Please login to your online account to display your discounted pricing

Rabbit anti-HNF-3 alpha/FoxA1, Clone: SP88, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP21249301ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP21249301

  • / Each
 

Description & Specifications

Specifications

Antigen HNF-3 alpha/FoxA1
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Clone SP88
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation PBS/1% BSA buffer pH 7.6
Host Species Rabbit
Immunogen Synthetic peptide derived from near C-terminus of human FOXA1 protein.
Isotype IgG
Purification Method Purified
Quantity 0.1mL
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3169
Test Specificity Predicted to cross react with Frog, Mouse, Rat.
Reconstitution Protein A purified