Please login to your online account to display your discounted pricing

Rabbit anti-HNF-3 alpha/FoxA1, Clone: SP88, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP21249305ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP21249305

  • / Each
 

Description & Specifications

Specifications

Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Cross Reactivity Human
Primary or Secondary Primary
Antigen HNF-3 alpha/FoxA1
Host Species Rabbit
Isotype IgG
Regulatory Status RUO
Formulation PBS/1% BSA buffer pH 7.6
Monoclonal or Polyclonal Monoclonal
Quantity 0.5mL
Conjugate Unlabeled
Clone SP88
Format Purified
Immunogen Synthetic peptide derived from near C-terminus of human FOXA1 protein.
Purification Method Purified
Gene ID (Entrez) 3169
Storage Requirements Store at 4°C. Do not freeze.
Reconstitution Protein A purified
Test Specificity Predicted to cross react with Frog, Mouse, Rat.