Please login to your online account to display your discounted pricing

Rabbit anti-Glypican 3, Clone: SP86, Novus Biologicals™

$309.05 - $551.93

Specifications

Antigen Glypican 3
Clone SP86
Immunogen Synthetic peptide derived from C-terminus of human glypican-3 protein.
Gene ID (Entrez) 2719
Formulation PBS/1% BSA buffer pH 7.6
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP21249101

 
novus biologicals canada
NBP21249101ML
0.1mL Each for $309.05

NBP21249105

 
novus biologicals canada
NBP21249105ML
0.5mL Each for $551.93
Description & Specifications

Specifications

Antigen Glypican 3
Clone SP86
Immunogen Synthetic peptide derived from C-terminus of human glypican-3 protein.
Gene ID (Entrez) 2719
Formulation PBS/1% BSA buffer pH 7.6
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rabbit
Isotype IgG
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blot