Please login to your online account to display your discounted pricing

Rabbit anti-Glypican 3, Clone: SP86, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP21249105ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP21249105

  • / Each
 

Description & Specifications

Specifications

Formulation PBS/1% BSA buffer pH 7.6
Host Species Rabbit
Immunogen Synthetic peptide derived from C-terminus of human glypican-3 protein.
Quantity 0.5mL
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Test Specificity Predicted to cross react with Frog.
Cross Reactivity Human
Antigen Glypican 3
Format Purified
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Isotype IgG
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Gene ID (Entrez) 2719
Clone SP86
Conjugate Unlabeled
Purification Method Purified
Reconstitution Protein A purified