Please login to your online account to display your discounted pricing

Rabbit anti-Glypican 3, Clone: SP86, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP21249101ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP21249101

  • / Each
 

Description & Specifications

Specifications

Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Primary or Secondary Primary
Host Species Rabbit
Cross Reactivity Human
Gene ID (Entrez) 2719
Formulation PBS/1% BSA buffer pH 7.6
Immunogen Synthetic peptide derived from C-terminus of human glypican-3 protein.
Isotype IgG
Monoclonal or Polyclonal Monoclonal
Conjugate Unlabeled
Purification Method Purified
Clone SP86
Antigen Glypican 3
Test Specificity Predicted to cross react with Frog.
Format Purified
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Quantity 0.1mL
Reconstitution Protein A purified