Please login to your online account to display your discounted pricing

Rabbit anti-F4/80/EMR1, Clone: SP115, Novus Biologicals™

$362.00 - $549.00

Specifications

Antigen F4/80/EMR1
Clone SP115
Immunogen Synthetic peptide derived from N-terminus of mouse F4/80 protein.
Gene ID (Entrez) 13733
Formulation PBS with 1% BSA, pH 7.6
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP21250605

 
novus biologicals canada
NBP21250605ML
0.5mL Each for $549.00

NBP21250601

 
novus biologicals canada
NBP21250601ML
0.1mL Each for $362.00
Description & Specifications

Specifications

Antigen F4/80/EMR1
Clone SP115
Immunogen Synthetic peptide derived from N-terminus of mouse F4/80 protein.
Gene ID (Entrez) 13733
Formulation PBS with 1% BSA, pH 7.6
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rabbit
Cross Reactivity Human
Cross Reactivity Mouse
Isotype IgG
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)