Please login to your online account to display your discounted pricing

Rabbit anti-F4/80/EMR1, Clone: SP115, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP21250605ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP21250605

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Cross Reactivity Mouse
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Test Specificity Predicted to cross react with Rat.
Antigen F4/80/EMR1
Immunogen Synthetic peptide derived from N-terminus of mouse F4/80 protein.
Regulatory Status RUO
Quantity 0.5mL
Monoclonal or Polyclonal Monoclonal
Format Purified
Conjugate Unlabeled
Gene ID (Entrez) 13733
Host Species Rabbit
Clone SP115
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Formulation PBS with 1% BSA, pH 7.6
Purification Method Purified
Isotype IgG
Reconstitution Protein A purified