Please login to your online account to display your discounted pricing

Rabbit anti-F4/80/EMR1, Clone: SP115, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP21250601ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP21250601

  • / Each
 

Description & Specifications

Specifications

Formulation PBS with 1% BSA, pH 7.6
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Immunogen Synthetic peptide derived from N-terminus of mouse F4/80 protein.
Monoclonal or Polyclonal Monoclonal
Antigen F4/80/EMR1
Format Purified
Host Species Rabbit
Isotype IgG
Purification Method Purified
Quantity 0.1mL
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Gene ID (Entrez) 13733
Reconstitution Protein A purified
Clone SP115
Conjugate Unlabeled
Test Specificity Predicted to cross react with Rat.
Cross Reactivity Human
Cross Reactivity Mouse