Please login to your online account to display your discounted pricing

Rabbit anti-ErbB3/Her3, Clone: SP71, Novus Biologicals™

$309.05 - $551.93

Specifications

Antigen ErbB3/Her3
Clone SP71
Immunogen Synthetic peptide corresponding to internal region of human ErbB3 Antibody (SP71).
Gene ID (Entrez) 2065
Formulation PBS/1% BSA buffer pH 7.6
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP21248001

 
novus biologicals canada
NBP21248001ML
0.1mL Each for $309.05

NBP21248005

 
novus biologicals canada
NBP21248005ML
0.5mL Each for $551.93
Description & Specifications

Specifications

Antigen ErbB3/Her3
Clone SP71
Immunogen Synthetic peptide corresponding to internal region of human ErbB3 Antibody (SP71).
Gene ID (Entrez) 2065
Formulation PBS/1% BSA buffer pH 7.6
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rabbit
Cross Reactivity Human
Isotype IgG
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)