Please login to your online account to display your discounted pricing

Rabbit anti-ErbB3/Her3, Clone: SP71, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP21248005ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP21248005

  • / Each
 

Description & Specifications

Specifications

Antigen ErbB3/Her3
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Quantity 0.5mL
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Conjugate Unlabeled
Format Purified
Formulation PBS/1% BSA buffer pH 7.6
Immunogen Synthetic peptide corresponding to internal region of human ErbB3 Antibody (SP71).
Isotype IgG
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Reconstitution Protein A purified
Cross Reactivity Human
Host Species Rabbit
Purification Method Purified
Gene ID (Entrez) 2065
Test Specificity Predicted cross-reactivity based on sequence identity: Mouse, Rat.
Clone SP71