Please login to your online account to display your discounted pricing

Rabbit anti-ErbB3/Her3, Clone: SP71, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP21248001ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP21248001

  • / Each
 

Description & Specifications

Specifications

Antigen ErbB3/Her3
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Clone SP71
Conjugate Unlabeled
Format Purified
Formulation PBS/1% BSA buffer pH 7.6
Host Species Rabbit
Immunogen Synthetic peptide corresponding to internal region of human ErbB3 Antibody (SP71).
Isotype IgG
Purification Method Purified
Quantity 0.1mL
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 2065
Test Specificity Predicted cross-reactivity based on sequence identity: Mouse, Rat.
Reconstitution Protein A purified
Target Species Human