Please login to your online account to display your discounted pricing

Rabbit anti-EGF R/ErbB1, Clone: SP9, Novus Biologicals™

$309.05 - $368.59

Specifications

Antigen EGF R/ErbB1
Clone SP9
Immunogen Synthetic peptide from C-terminus of human EGFR protein.
Gene ID (Entrez) 1956
Formulation TBS/1% BSA buffer pH 7.5. Tissue culture supernatant.
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP21246801

 
novus biologicals canada
NBP21246801ML
0.1mL Each for $309.05

NBP21246805

 
novus biologicals canada
NBP21246805ML
0.5mL Each for $368.59
Description & Specifications

Specifications

Antigen EGF R/ErbB1
Clone SP9
Immunogen Synthetic peptide from C-terminus of human EGFR protein.
Gene ID (Entrez) 1956
Formulation TBS/1% BSA buffer pH 7.5. Tissue culture supernatant.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rabbit
Isotype IgG
Conjugate Unlabeled
Format Supernatant
Purification Method Supernatant
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)