Please login to your online account to display your discounted pricing

Rabbit anti-EGF R/ErbB1, Clone: SP9, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP21246801ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP21246801

  • / Each
 

Description & Specifications

Specifications

Format Supernatant
Formulation TBS/1% BSA buffer pH 7.5. Tissue culture supernatant.
Host Species Rabbit
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Immunogen Synthetic peptide from C-terminus of human EGFR protein.
Quantity 0.1mL
Conjugate Unlabeled
Cross Reactivity Human
Antigen EGF R/ErbB1
Isotype IgG
Purification Method Supernatant
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Test Specificity Predicted to cross react with Bovine, Mouse, Porcine, Rat.
Clone SP9
Gene ID (Entrez) 1956