Please login to your online account to display your discounted pricing

Rabbit anti-EGF R/ErbB1, Clone: SP84, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP21248905ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP21248905

  • / Each
 

Description & Specifications

Specifications

Host Species Rabbit
Cross Reactivity Human
Storage Requirements Store at 4°C. Do not freeze.
Antigen EGF R/ErbB1
Regulatory Status RUO
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Gene ID (Entrez) 1956
Immunogen Synthetic peptide corresponding to C-terminus of human EGFR protein.
Format Purified
Monoclonal or Polyclonal Monoclonal
Clone SP84
Reconstitution Protein A purified
Purification Method Purified
Conjugate Unlabeled
Formulation PBS/1% BSA buffer pH 7.6
Isotype IgG
Primary or Secondary Primary
Test Specificity Predicted to cross react with Bovine, Canine, Frog, Mouse, Porcine, Rat .
Quantity 0.5mL