Please login to your online account to display your discounted pricing

Rabbit anti-Desmin, Clone: SP138, Novus Biologicals™

$309.05 - $552.33

Specifications

Antigen Desmin
Clone SP138
Immunogen Synthetic peptide derived from the C-terminus of human desmin protein.
Gene ID (Entrez) 1674
Formulation PBS/1% BSA buffer pH 7.6
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP21251205

 
novus biologicals canada
NBP21251205ML
0.5mL Each for $552.33

NBP21251201

 
novus biologicals canada
NBP21251201ML
0.1mL Each for $309.05
Description & Specifications

Specifications

Antigen Desmin
Clone SP138
Immunogen Synthetic peptide derived from the C-terminus of human desmin protein.
Gene ID (Entrez) 1674
Formulation PBS/1% BSA buffer pH 7.6
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rabbit
Cross Reactivity Human
Isotype IgG
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting