Please login to your online account to display your discounted pricing

Rabbit anti-Desmin, Clone: SP138, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP21251205ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP21251205

  • / Each
 

Description & Specifications

Specifications

Host Species Rabbit
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Cross Reactivity Human
Antigen Desmin
Format Purified
Isotype IgG
Purification Method Purified
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Gene ID (Entrez) 1674
Test Specificity Predicted to cross react with Bovine, Chicken, Canine, Frog, Hamster, Mouse, Porcine, Rat.
Reconstitution Protein A purified
Clone SP138
Conjugate Unlabeled
Formulation PBS/1% BSA buffer pH 7.6
Immunogen Synthetic peptide derived from the C-terminus of human desmin protein.
Quantity 0.5mL
Monoclonal or Polyclonal Monoclonal