Please login to your online account to display your discounted pricing

Rabbit anti-Desmin, Clone: SP138, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP21251201ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP21251201

  • / Each
 

Description & Specifications

Specifications

Host Species Rabbit
Quantity 0.1mL
Formulation PBS/1% BSA buffer pH 7.6
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Immunogen Synthetic peptide derived from the C-terminus of human desmin protein.
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Predicted to cross react with Bovine, Chicken, Canine, Frog, Hamster, Mouse, Porcine, Rat.
Conjugate Unlabeled
Cross Reactivity Human
Antigen Desmin
Format Purified
Storage Requirements Store at 4°C. Do not freeze.
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 1674
Clone SP138
Isotype IgG
Purification Method Purified
Reconstitution Protein A purified