Please login to your online account to display your discounted pricing

Rabbit anti-Cytokeratin 8, Clone: SP102, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP21250001ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP21250001

  • / Each
 

Description & Specifications

Specifications

Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Quantity 0.1mL
Storage Requirements Store at 4°C. Do not freeze.
Monoclonal or Polyclonal Monoclonal
Host Species Rabbit
Gene ID (Entrez) 3856
Formulation PBS/1% BSA buffer pH 7.6
Reconstitution Protein A purified
Antigen Cytokeratin 8
Cross Reactivity Human
Conjugate Unlabeled
Purification Method Purified
Regulatory Status RUO
Test Specificity Predicted to cross react with Bovine, Mouse, Porcine, Rat.
Immunogen Synthetic peptide corresponding to C-terminus of human keratin 8.
Primary or Secondary Primary
Isotype IgG
Clone SP102
Format Purified