Please login to your online account to display your discounted pricing

Rabbit anti-Cytokeratin 8, Clone: SP102, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP21250005ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP21250005

  • / Each
 

Description & Specifications

Specifications

Formulation PBS/1% BSA buffer pH 7.6
Isotype IgG
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Clone SP102
Antigen Cytokeratin 8
Format Purified
Host Species Rabbit
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Immunogen Synthetic peptide corresponding to C-terminus of human keratin 8.
Purification Method Purified
Quantity 0.5mL
Primary or Secondary Primary
Gene ID (Entrez) 3856
Test Specificity Predicted to cross react with Bovine, Mouse, Porcine, Rat.
Reconstitution Protein A purified
Conjugate Unlabeled
Cross Reactivity Human
Monoclonal or Polyclonal Monoclonal