missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Rabbit anti-Cytokeratin 7, Clone: R17-S, Novus Biologicals™

$309.05 - $309.05

Specifications

Antigen Cytokeratin 7
Clone R17-S
Immunogen Peptide derived from N-terminal sequence of human cytokeratin 7.
Gene Accession No. P08729
Gene ID (Entrez) 3855
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP13015201 Novus Biologicals Canada
NBP13015201ML
0.1mL Each for $309.05
Specifications Description & Specifications

Specifications

Cytokeratin 7
Peptide derived from N-terminal sequence of human cytokeratin 7.
3855
Monoclonal
Store at 4°C. Do not freeze.
IgG
Affinity Purified
Immunocytochemistry,Immunofluorescence,Immunohistochemistry,Immunohistochemistry (Frozen),Immunohistochemistry (Paraffin)
R17-S
P08729
20mM Tris-HCl, pH 8.0 with 20mg/mL BSA.
Primary
Rabbit
Unlabeled
Affinity Purified