Please login to your online account to display your discounted pricing

Rabbit anti-Cytokeratin 7, Clone: R17-S, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP13015201ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP13015201

  • / Each
 

Description & Specifications

Specifications

Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Host Species Rabbit
Conjugate Unlabeled
Cross Reactivity Human
Isotype IgG
Clone R17-S
Monoclonal or Polyclonal Monoclonal
Regulatory Status RUO
Gene ID (Entrez) 3855
Antigen Cytokeratin 7
Purification Method Affinity Purified
Test Specificity This antibody is specific for the N terminus o Cytokeratin 7
Quantity 0.1mL
Primary or Secondary Primary
Gene Accession No. P08729
Immunogen Peptide derived from N-terminal sequence of human cytokeratin 7.
Formulation 20mM Tris-HCl, pH 8.0 with 20mg/mL BSA.
Format Affinity Purified
Storage Requirements Store at 4°C. Do not freeze.