Please login to your online account to display your discounted pricing

Rabbit anti-Cytokeratin 7, Clone: R17-S, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP13015210ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP13015210

  • / Each
 

Description & Specifications

Specifications

Format Affinity Purified
Formulation 20mM Tris-HCl, pH 8.0 with 20mg/mL BSA.
Host Species Rabbit
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Immunogen Peptide derived from N-terminal sequence of human cytokeratin 7.
Primary or Secondary Primary
Conjugate Unlabeled
Antigen Cytokeratin 7
Gene Accession No. P08729
Isotype IgG
Purification Method Affinity Purified
Quantity 1mL
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3855
Test Specificity This antibody is specific for the N terminus o Cytokeratin 7
Cross Reactivity Human
Clone R17-S