Please login to your online account to display your discounted pricing

Rabbit anti-Cytokeratin 6, Clone: SP87, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP21249205ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP21249205

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Monoclonal or Polyclonal Monoclonal
Format Purified
Immunogen Synthetic peptide derived from C-terminus of human keratin 6 protein.
Host Species Rabbit
Primary or Secondary Primary
Regulatory Status RUO
Gene ID (Entrez) 3853
Quantity 0.5mL
Purification Method Purified
Formulation PBS/1% BSA buffer pH 7.6
Isotype IgG
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Conjugate Unlabeled
Storage Requirements Store at 4°C. Do not freeze.
Antigen Cytokeratin 6
Reconstitution Protein A purified
Clone SP87
Test Specificity Predicted to cross react with Bovine.