Please login to your online account to display your discounted pricing

Rabbit anti-Cytokeratin 6, Clone: SP87, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP21249201ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP21249201

  • / Each
 

Description & Specifications

Specifications

Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Immunogen Synthetic peptide derived from C-terminus of human keratin 6 protein.
Isotype IgG
Quantity 0.1mL
Regulatory Status RUO
Gene ID (Entrez) 3853
Conjugate Unlabeled
Antigen Cytokeratin 6
Format Purified
Formulation PBS/1% BSA buffer pH 7.6
Host Species Rabbit
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Clone SP87
Cross Reactivity Human
Purification Method Purified
Test Specificity Predicted to cross react with Bovine.
Reconstitution Protein A purified