Please login to your online account to display your discounted pricing

Rabbit anti-Cytokeratin 5, Clone: SP27, Novus Biologicals™

$309.33 - $518.93

Specifications

Antigen Cytokeratin 5
Clone SP27
Immunogen Synthetic peptide from C-terminus of human cytokeratin 5.
Gene ID (Entrez) 3852
Formulation TBS/1% BSA buffer pH 7.5
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP21247301

 
novus biologicals canada
NBP21247301ML
0.1mL Each for $309.33

NBP21247305

 
novus biologicals canada
NBP21247305ML
0.5mL Each for $518.93
Description & Specifications

Specifications

Antigen Cytokeratin 5
Clone SP27
Immunogen Synthetic peptide from C-terminus of human cytokeratin 5.
Gene ID (Entrez) 3852
Formulation TBS/1% BSA buffer pH 7.5
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rabbit
Cross Reactivity Human
Isotype IgG
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting